Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAG3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 8 publications
$268.00 - $499.50
Specifications
Antigen | BAG3 |
---|---|
Dilution | Western Blot 1:1000-1:2000∼, Simple Western 1:100, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000∼, Immunohistochemistry-Frozen 1:1000-1:5000 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227398SS
![]() |
Novus Biologicals
NBP227398SS |
0.025 mL |
Each for $268.00
|
|
|||||
NBP227398
![]() |
Novus Biologicals
NBP227398 |
0.05 mL |
Each for $499.50
|
|
|||||
Description
BAG3 Polyclonal specifically detects BAG3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
BAG3 | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O95817 | |
9531 | |
A recombinant protein fragment corresponding to the C-terminal 196 amino acids of human BAG-3. Bag-3 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:1000-1:2000∼, Simple Western 1:100, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000∼, Immunohistochemistry-Frozen 1:1000-1:5000 | |
Polyclonal | |
Antisera | |
RUO | |
50 ul neat whole antisera with 0.05% Sodium Azide | |
BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307 | |
BAG3 | |
IgG | |
Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title