Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAG3 Antibody (V65P1E8*D2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP20112A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20112A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20112A30 Supplier Novus Biologicals Supplier No. NBP250112AF350
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BAG3 Monoclonal specifically detects BAG3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen BAG3
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V65P1E8*D2
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307
Gene Symbols BAG3
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide MTDTPGNPAAP
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9531.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.