Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578292 Supplier Invitrogen™ Supplier No. PA578292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: U87-MG, MCF-7, rat heart Predicted Reactivity: Mouse (85%), Rhesus Monkey (97%), Bovine (83%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

BAG3 are among BAG proteins that compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. All the BAG proteins have an approximately 45-amino acid BAG domain near the C terminus but differ markedly in their N-terminal regions. The protein encoded by this gene contains a WW domain in the N-terminal region and a BAG domain in the C-terminal region. The BAG domains of BAG1, BAG2, and BAG3 interact specifically with the Hsc70 ATPase domain in vitro and in mammalian cells. All 3 proteins bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAG3
Applications Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene BAG3
Gene Accession No. O95817
Gene Alias AA407278; BAG family molecular chaperone regulator 3; BAG3; BAG-3; BCL2 associated athanogene 3; Bcl-2-associated athanogene 3; Bcl2-associated athanogene 3; BCL2-binding athanogene 3; bcl-2-binding protein Bis; Bcl-2-interacting death suppressor; BIS; CAIR-1; DKFZp434E0610; docking protein CAIR-1; MFM6; mg638; MGC104307; MNCb-2243
Gene Symbols BAG3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BAG3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293524, 9531
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.