Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAG4 Monoclonal Antibody (2G9)
GREENER_CHOICE

Catalog No. PIMA520758
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520758 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520758 Supplier Invitrogen™ Supplier No. MA520758
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MSALRRSGYG PSDGPSYGRY YGPGGGDVPV HPPPPLYPLR PEPPQPPISW RVRGGGPAET TWLGEGGGGD GYYPSGGAWP EPGRAGGSHQ EQPPYPSYNS NYWNSTARSR APYPSTYPVR PELQGQSLNS YTNGAYGPTY PPGPGANTAS YSGAYYAPGY TQTSYSTEVP STYRSSGNSP TPVSRWIYPQ QDCQTEAPPL RGQVPGYPPS QNPGMTLPHY PYGDGNRSVP QSGPTVRPQE DAWASPGAYG MGGRYPWPSS APSAPPGNLY MTESTSPWPS SGSPQSPPSP PVQQPKDSSY PYSQSDQSMN RHNFPCSVHQ YESSGTVNND DSDLLDSQVQ YSAEPQLYGN ATSDHPNNQD QSSSLPEECV PSDESTPPSI KKIIHVLEKV QYLEQEVEEF VGKKTDKAYW LLEEMLTKEL LELDSVETGG QDSVRQARKE AVCKIQAILE KLEKKGL.

The protein encoded by this gene is a member of the BAG1-related protein family. BAG1 is an anti-apoptotic protein that functions through interactions with a variety of cell apoptosis and growth related proteins including BCL-2, Raf-protein kinase, steroid hormone receptors, growth factor receptors and members of the heat shock protein 70 kDa family. This protein contains a BAG domain near the C-terminus, which could bind and inhibit the chaperone activity of Hsc70/Hsp70. This protein was found to be associated with the death domain of tumor necrosis factor receptor type 1 (TNF-R1) and death receptor-3 (DR3), and thereby negatively regulates downstream cell death signaling. The regulatory role of this protein in cell death was demonstrated in epithelial cells which undergo apoptosis while integrin mediated matrix contacts are lost.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAG4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene BAG4
Gene Accession No. O95429
Gene Alias 2410112I15Rik; BAG cochaperone 4; BAG family molecular chaperone regulator 4; BAG4; BAG-4; BAG-family molecular chaperone regulator-4; BCL2 associated athanogene 4; Bcl-2-associated athanogene 4; BCL2-associated athanogene 4; silencer of death domains; SODD
Gene Symbols BAG4
Host Species Mouse
Immunogen BAG4 (AAH38505, 1 a.a. ~ 457 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9530
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.