Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BAG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109441
Description
Immunogen sequence: MSALRRSGYG PSDGPSYGRY YGPGGGDVPV HPPPPLYPLR PEPPQPPISW RVRGGGPAET TWLGEGGGGD GYYPSGGAWP EPGRAGGSHQ SLNSYTNGAY.
The protein encoded by this gene is a member of the BAG1-related protein family. BAG1 is an anti-apoptotic protein that functions through interactions with a variety of cell apoptosis and growth related proteins including BCL-2, Raf-protein kinase, steroid hormone receptors, growth factor receptors and members of the heat shock protein 70 kDa family. This protein contains a BAG domain near the C-terminus, which could bind and inhibit the chaperone activity of Hsc70/Hsp70. This protein was found to be associated with the death domain of tumor necrosis factor receptor type 1 (TNF-R1) and death receptor-3 (DR3), and thereby negatively regulates downstream cell death signaling. The regulatory role of this protein in cell death was demonstrated in epithelial cells which undergo apoptosis while integrin mediated matrix contacts are lost.
Specifications
| BAG4 | |
| Polyclonal | |
| Unconjugated | |
| BAG4 | |
| 2410112I15Rik; BAG cochaperone 4; BAG family molecular chaperone regulator 4; BAG4; BAG-4; BAG-family molecular chaperone regulator-4; BCL2 associated athanogene 4; Bcl-2-associated athanogene 4; BCL2-associated athanogene 4; silencer of death domains; SODD | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 361167, 67384, 9530 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.06 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O95429, Q8CI61 | |
| BAG4 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human BAG4 (NP_001191807.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction