Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAG5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596429 Supplier Invitrogen™ Supplier No. PA596429
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVLIALLMGV NNNETCRHLS CVLSGLIADL DALDVCGRTE IRNYRREVVE DINKLLKYLD LEEEADTTKA FDLRQNHSIL KIEKVLKRMR EIKNELLQAQ NPSELYLSSK TELQGLIGQL DEVSLEKNPC IREARRRAVI EVQTLITYID LKEALEKRKL FACEEHPSHK AVWNVLGNLS EIQGEVLSFD GNRTDKNYIR LEELLTKQLL ALDAVDPQGE EKCKAARKQA VRLAQNILSY LDLKSDEWEY; Positive Samples: SH-SY5Y, MCF7, Mouse lung, Mouse testis, Rat brain, Rat lung.

It has been hypothesizes that the BAG-5 protein will induce the death of nigral neurons through its predicted interaction with hsp70, which will cause increased protein aggregation and cell death by disinhibition of hsp70's anti-apoptotic function. It is believed that BAG-5 will play an important role in the mechanisms of neuronal death. BAG-5 may also be of interest due to its possible role as a modulator of the hsp70/hsp40 chaperone axis or its possible interaction and coordination of localization/ modulation of other BAG containing proteins via BAG-BAG heterodimerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAG5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BAG5
Gene Accession No. Q5QJC9, Q8CI32, Q9UL15
Gene Alias 1600025G07Rik; 4930405J06Rik; BAG cochaperone 5; BAG family molecular chaperone regulator 5; Bag5; BAG-5; BAG-family molecular chaperone regulator-5; BAG-family molecular chaperone regulator-5 (BAG-5); BCL2 associated athanogene 5; Bcl-2-associated athanogene 5; BCL2-associated athanogene 5; KIAA0873; LOC522854 protein
Gene Symbols BAG5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 198-447 of human BAG5 (NP_0048641).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366734, 70369, 9529
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.