Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BAG5 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142006
This item is not returnable.
View return policy
Description
Additional Information: Variants (NP_004864.1; NP_001015048.1) encode the same isoform. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
It has been hypothesizes that the BAG-5 protein will induce the death of nigral neurons through its predicted interaction with hsp70, which will cause increased protein aggregation and cell death by disinhibition of hsp70's anti-apoptotic function. It is believed that BAG-5 will play an important role in the mechanisms of neuronal death. BAG-5 may also be of interest due to its possible role as a modulator of the hsp70/hsp40 chaperone axis or its possible interaction and coordination of localization/ modulation of other BAG containing proteins via BAG-BAG heterodimerization.
Specifications
| BAG5 | |
| Polyclonal | |
| Unconjugated | |
| BAG5 | |
| 1600025G07Rik; 4930405J06Rik; BAG cochaperone 5; BAG family molecular chaperone regulator 5; Bag5; BAG-5; BAG-family molecular chaperone regulator-5; BAG-family molecular chaperone regulator-5 (BAG-5); BCL2 associated athanogene 5; Bcl-2-associated athanogene 5; BCL2-associated athanogene 5; KIAA0873; LOC522854 protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 9529 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UL15 | |
| BAG5 | |
| Peptide with sequence DMGNQHPSISRLQ-C (aa 2-14). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction