Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAG6 Mouse anti-Human, Mouse, Rat, Clone: 1B8D3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | BAG6 |
|---|---|
| Clone | 1B8D3 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7079
![]()
|
Proteintech Group Inc
666611IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a protein with two leucine zipper domains, a ribosomal protein S14 signature domain and a Sec39 like domain. The protein is thought to be involved in Golgi-to-ER transport. Mutations in this gene are associated with short stature, optic nerve atrophy, and Pelger-Huet anomaly. Involved in Golgi-to-endoplasmic reticulum (ER) retrograde tranport; the function is proposed to depend on its association in the NRZ complex which is believed to play a role in SNARE assembly at the ER.Specifications
| BAG6 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| BAG 6, BAG6, BAT3, D6S52E, G3, HLA B associated transcript 3, Protein G3 | |
| BAG6 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1B8D3 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P46379, Q6MG49, Q9Z1R2 | |
| 224727, 7917, 94342 | |
| BAG6 Fusion Protein Ag24525 | |
| Primary | |
| -20°C | |
| BAG6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title