Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAG6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562770 Supplier Invitrogen™ Supplier No. PA562770
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDAYLSGMPA KRRKTMQGEG PQLLLSEAVS RAAKAAGARP LTSPESLSRD LEAPEVQESY RQQLRSDIQK RLQEDPNYS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAG6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Bag6
Gene Accession No. P46379
Gene Alias 2410045D21Rik; AA408914; BAG 6; BAG family molecular chaperone regulator 6; Bag6; BAG-6; BAT 3; BAT3; BCL2 associated athanogene 6; BCL2-associated athanogene 6; D17H6S52E; D6S52E; DAQB-195H10.3; G3; HLA-B associated transcript 3; HLA-B associated transcript-3; HLA-B-associated transcript 3; Large proline-rich protein BAG6; large proline-rich protein BAT3; protein G3; Protein Scythe; scythe
Gene Symbols Bag6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BAG6. Recombinant protein control fragment (Product #RP-103149).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7917
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.