Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAGE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587687 Supplier Invitrogen™ Supplier No. PA587687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLMKEESPVV SWRLEPEDGT ALDVHFVSTL EPLSNAVKRN VPRCIIILVL QEPTPFRISV TSSCFVQNTL TKLLKDRRKM QTVQCATARE TS; Positive Samples: A-431.

BAGE2 (BAGE Family Member 2) is a Protein Coding gene. Candidate gene encoding tumor antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAGE2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BAGE2
Gene Accession No. Q86Y30
Gene Alias B melanoma antigen 2; B melanoma antigen family member 2; B melanoma antigen family, member 2; BAGE2; cancer/testis antigen 2.2; cancer/testis antigen family 2, member 2; CT2.2
Gene Symbols BAGE2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 18-109 of human BAGE2 (NP_8722882).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85319
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.