Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAGE4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988832
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8832 Supplier Bioss Supplier No. BS9116R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BAGE4, also named as MLL3P and CT2.4, belongs to the BAGE family. It is a candidate gene encoding tumor antigens. BAGE4 is Cancer/testis antigen 2.4.

Specifications

Antigen BAGE4
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BAGE4
Gene Accession No. Q86Y28
Gene Alias B melanoma antigen 4; B melanoma antigen family member 4; B melanoma antigen family, member 4; BAGE4; Cancer/testis antigen 2.4; cancer/testis antigen family 2, member 4; CT2.4; MLL3 pseudogene; MLL3P
Gene Symbols BAGE4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BAGE4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85317
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.