Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAH-1 Mouse anti-Human, Brilliant Violet 421, Clone: BAH-1 (also known as BAH1), BD Optibuild™

Catalog No. BDB743955 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743955 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743955 Supplier BD Biosciences Supplier No. 743955
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The BAH-1 monoclonal antibody was raised against human megakaryocytic cells. It was originally proposed that clone BAH-1 was specific for human CD110, also known as c-Mpl or TPO-R. Data has emerged to suggest that BAH-1 antibody may not recognize CD110. However, there is strong evidence that it recognizes an antigen that is highly expressed on human Megakaryocyte-Erythroid Progenitor (MEP) cells. In combination with CD71, BAH-1 expression has been used to distinguish subsets of Hematopoietic Stem Cells and Myeloid Progenitors in human fetal liver, cord blood, and adult bone marrow. BAH-1 antibody can induce megakaryocytopoiesis of both mouse and human bone marrow cells in vitro.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAH-1
Classification Monoclonal
Clone BAH-1 (also known as BAH1)
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias BAH1
Host Species Mouse
Immunogen Human Megakaryocytic Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.