Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAH-1 Mouse, anti-Human, R718, Clone: BAH-1, BD OptiBuild™

Catalog No. BDB752213 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752213 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752213 Supplier BD Biosciences Supplier No. 752213
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The BAH-1 monoclonal antibody was raised against human megakaryocytic cells. It was originally proposed that clone BAH-1 was specific for human CD110, also known as c-Mpl or TPO-R. Data has emerged to suggest that BAH-1 antibody may not recognize CD110. However, there is strong evidence that it recognizes an antigen that is highly expressed on human Megakaryocyte-Erythroid Progenitor (MEP) cells. In combination with CD71, BAH-1 expression has been used to distinguish subsets of Hematopoietic Stem Cells and Myeloid Progenitors in human fetal liver, cord blood, and adult bone marrow. BAH-1 antibody can induce megakaryocytopoiesis of both mouse and human bone marrow cells in vitro.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BAH-1
Applications Flow Cytometry
Classification Monoclonal
Clone BAH-1
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.