Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAHCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585013 Supplier Invitrogen™ Supplier No. PA585013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGLDKSGYFE LPTSSQDCAR PGHQDPLGGK APQACCTLDK TVGKEAPAGP PGAQKVARIR HQQHLMAAEV EQGGIGAEAK RKSLELA Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 82%.

BAHCC1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAHCC1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BAHCC1
Gene Accession No. Q9P281
Gene Alias BAH and coiled-coil domain-containing protein 1; BAH domain and coiled-coil containing 1; BAH domain-containing protein 2; BAHCC1; BAHD2; Bromo adjacent homology domain-containing protein 2; KIAA1447
Gene Symbols BAHCC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BAHCC1. Recombinant protein control fragment (Product #RP-108101).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57597
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.