Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAI1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 4 publications

$207.00 - $499.50

Specifications

Antigen BAI1
Concentration 1 mg/mL
Dilution Western Blot reported in scientific literature, Immunohistochemistry 1:1000-1:3000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000-1:3000
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11081586T
SDP
View Documents
Novus Biologicals
NB11081586SS
0.025 mL
Each for $207.00
Only null left
Add to Cart
 
NB11081586
SDP
View Documents
Novus Biologicals
NB11081586
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

BAI1 Polyclonal specifically detects BAI1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

BAI1
Western Blot reported in scientific literature, Immunohistochemistry 1:1000-1:3000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000-1:3000
Polyclonal
Rabbit
Alzheimers Research, Angiogenesis, Apoptosis, Cancer, GPCR, Neurodegeneration, Neuroscience
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF
BAI1
IgG
Affinity Purified
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse
O14514
575
Synthetic peptide made to a C-terminal portion of the human BAI1 protein (within residues 1500-1584). [Swiss-Prot# O14514]
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.