Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAI1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N1108186A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1108186A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1108186A30 Supplier Novus Biologicals Supplier No. NB11081586AF350
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BAI1 Polyclonal specifically detects BAI1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen BAI1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF
Gene Symbols ADGRB1
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human BAI1 protein (within residues 1500-1584). [Swiss-ProtNo. O14514]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, Angiogenesis, Apoptosis, Cancer, GPCR, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 575.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.