Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575348 Supplier Thermo Scientific Supplier No. PA575348
Add to Cart
Add to Cart

BAI1 Polyclonal Antibody

BAI1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen BAI1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ADGRB1
Gene Accession No. C0HL12, O14514, Q3UHD1
Gene Alias ADGRB1; Adhesion G protein-coupled receptor B1; B830018M07Rik; Bai1; brain-specific angiogenesis inhibitor 1; FLJ41988; GDAIF; mKIAA4089; R75078; Vasculostatin-120; Vasculostatin-40; Vstat120; Vstat40; Vstat-40
Gene Symbols ADGRB1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 711-760 of Human BAI1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107831, 362931, 575
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.