Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    BAI2 Antibody (2528B), mFluor Violet 500 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB9338MFV500
Description
BAI2 Monoclonal antibody specifically detects BAI2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| BAI2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Angiogenesis, Cancer, GPCR, Neuroscience | |
| 576 | |
| Store at 4C in the dark. | 
| Western Blot, Flow Cytometry, CyTOF | |
| 2528B | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| brain-specific angiogenesis inhibitor 2, Brain-specific angiongenesis inhibitor-2 | |
| Human embryonic kidney cell, HEK293-derived human BAI2, Phe21-Asp911, Accession # O60241-3 | |
| RUO | |
| Primary | |
| Human | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            