Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BAI2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599883
Description
Antibody detects endogenous levels of total BAI2.
BAI1, a p53-target gene, encodes brain-specific angiogenesis inhibitor, a seven-span transmembrane protein and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities and may also play a role in angiogenesis.
Specifications
BAI2 | |
Polyclonal | |
Unconjugated | |
ADGRB2 | |
Adgrb2; Adhesion G protein-coupled receptor B2; BAI2; Brain-specific angiogenesis inhibitor 2; G protein-coupled receptor brain-specific angiogenesis inhibitor 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
230775, 576 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O60241, Q8CGM1 | |
ADGRB2 | |
A synthesized peptide derived from human ADGRB2(Accession O60241), corresponding to amino acid residues P1418-D1468. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction