Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111358
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111358 Supplier Invitrogen™ Supplier No. PA5111358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CLVGPEGSLS FSPLPGNILV PMAASPGLGE PPPPQEANPV YMCGEGGLRQ LDLTWLRPTE PGSEGDYMVL PRRTLSL Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 94%.

BAI1, a p53-target gene, encodes brain-specific angiogenesis inhibitor, a seven-span transmembrane protein and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities and may also play a role in angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAI2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADGRB2
Gene Accession No. O60241
Gene Alias Adgrb2; Adhesion G protein-coupled receptor B2; BAI2; Brain-specific angiogenesis inhibitor 2; G protein-coupled receptor brain-specific angiogenesis inhibitor 1
Gene Symbols ADGRB2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human BAI2. Recombinant protein control fragment (Product #RP-106402).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 576
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.