Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BAI2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533353
Description
This antibody is not recommended for use in ICC applications. Percent identity with other species by BLAST analysis: Human, Gorilla, Marmoset (100%) Orangutan, Monkey, Horse (95%) Hamster, Elephant, Panda, Pig (89%).
BAI1, a p53-target gene, encodes brain-specific angiogenesis inhibitor, a seven-span transmembrane protein and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities and may also play a role in angiogenesis.
Specifications
| BAI2 | |
| Polyclonal | |
| Unconjugated | |
| ADGRB2 | |
| Adgrb2; Adhesion G protein-coupled receptor B2; BAI2; Brain-specific angiogenesis inhibitor 2; G protein-coupled receptor brain-specific angiogenesis inhibitor 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 576 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O60241 | |
| ADGRB2 | |
| Synthetic 19 amino acid peptide from N-terminal extracellular domain of human BAI2. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction