Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAI3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533358
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533358 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533358 Supplier Invitrogen™ Supplier No. PA533358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Dog, Bat, Bovine, Horse, Rabbit, Opossum, Chicken (100%).

This p53-target gene encodes a brain-specific angiogenesis inhibitor, a seven-span transmembrane protein, and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities, and may also play a role in angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAI3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Adgrb3
Gene Accession No. O60242, Q80ZF8
Gene Alias A830096D10Rik; ADGRB3; Adhesion G protein-coupled receptor B3; BAI3; Brain-specific angiogenesis inhibitor 3; dJ91B17.1 (brain-specific angiogenesis inhibitor 3); G protein-coupled receptor BAI3; KIAA0550
Gene Symbols Adgrb3
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from C-terminus of human BAI3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100057127, 100348237, 210933, 301309, 421875, 481870, 577
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Horse, Chicken, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.