Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAI3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575813 Supplier Thermo Scientific Supplier No. PA575813
Add to Cart
Add to Cart

BAI3 Polyclonal Antibody

BAI3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen BAI3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Adgrb3
Gene Accession No. O60242, Q80ZF8
Gene Alias A830096D10Rik; ADGRB3; Adhesion G protein-coupled receptor B3; BAI3; Brain-specific angiogenesis inhibitor 3; dJ91B17.1 (brain-specific angiogenesis inhibitor 3); G protein-coupled receptor BAI3; KIAA0550
Gene Symbols Adgrb3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human BAI3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210933, 301309, 577
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.