Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAIAP2 Antibody (OTI 1D9), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271603PECY55
Description
BAIAP2 Monoclonal specifically detects BAIAP2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
BAIAP2 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
BAIAP2 | |
Full length recombinant protein of human BAIAP2 (NP_006331) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI 1D9 | |
Flow Cytometry | |
BAI1-associated protein 2IRS-58, BAI-associated protein 2, BAP2, brain-specific angiogenesis inhibitor 1-associated protein 2, Fas ligand-associated factor 3, FLAF3, Insulin receptor substrate p53, Insulin receptor substrate p53/p58, Insulin receptor substrate protein of 53 kDa, IRSp53, IRSp53/58, Protein BAP2 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
10458.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction