Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAIAP2L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121180
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121180 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121180 Supplier Invitrogen™ Supplier No. PA5121180
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HepG2, HeLa, 293T, A-549, Mouse lung. The target is usually found in the following locations: Cytoplasm, cytoskeleton. Immunogen sequence: DKHCGFANHI HYYHLQSAEL LNSKLPRWQE TCVDAIKVPE KIMNMIEEIK TPASTPVSGT PQASPMIERS NVVRKDYDTL SKCSPKMPPA PSGRAYTSPL IDMFNNPATA APNSQRVNNS TGTSEDPSLQ RSVSVATGLN MMKKQKVKTI FPHTAGSNKT LLSFAQGDVI TLLIPEEKDG WLYGEHDVSK A.

This gene encodes a member of the IMD (IRSp53/MIM homology domain) family. Members of this family can be subdivided in two groups, the IRSp53-like and MIM-like, based on the presence or absence of the SH3 (Src homology 3) domain. The protein encoded by this gene contains a conserved IMD, also known as F-actin bundling domain, at the N-terminus, and a canonical SH3 domain near the C-terminus, so it belongs to the IRSp53-like group. This protein is the substrate for insulin receptor tyrosine kinase and binds to the small GTPase Rac. It is involved in signal transduction pathways that link deformation of the plasma membrane and remodeling of the actin cytoskeleton. It also promotes actin assembly and membrane protrusions when overexpressed in mammalian cells, and is essential to the formation of a potent actin assembly complex during EHEC (Enterohemorrhagic Escherichia coli) pedestal formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAIAP2L1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BAIAP2L1
Gene Accession No. Q9DBJ3, Q9UHR4
Gene Alias 1300006M19Rik; AI585895; BAI1 associated protein 2 like 1; BAI1-associated protein 2-like 1; BAI1-associated protein 2-like protein 1; Baiap2l1; Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1; insulin receptor tyrosine kinase substrate; IRTKS; RGD1308452
Gene Symbols BAIAP2L1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 198-388 of human BAIAP2L1 (NP_061330.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55971, 66898
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.