Learn More
Invitrogen™ BAIAP2L2 Polyclonal Antibody

Description
Immunogen sequence: VVEVLVPEAQ NGWLYGKLEG SSASGWFPEA YVKALEEGPV NPMTPVTPMT SMTSMSPMTP MNPGNELPSR SYPLRGSHSL DDLLDRPGNS IAPSEYWDGQ SRSRTPSRVP SRAPSPAPPP LPSSRRSSMG STAVATDVKK LMSSEQYPP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.
Specifications
Specifications
| Antigen | BAIAP2L2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BAIAP2L2 |
| Gene Accession No. | Q6UXY1 |
| Gene Alias | BAI1 associated protein 2 like 2; BAI1-associated protein 2-like 2; BAI1-associated protein 2-like protein 2; Baiap2l2; BC048937; Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 2; pinkbar; Planar intestinal- and kidney-specific BAR domain protein; RGD1308786; UNQ9336/PRO34007 |
| Gene Symbols | BAIAP2L2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.