Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAIAP2L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101469 Supplier Invitrogen™ Supplier No. PA5101469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BAIAP2L2.

The protein encoded by this gene binds phosphoinositides and promotes the formation of planar or curved membrane structures. The encoded protein is found in RAB13-positive vesicles and at intercellular contacts with the plasma membrane. [provided by RefSeq, Dec 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAIAP2L2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BAIAP2L2
Gene Accession No. Q6UXY1, Q80Y61
Gene Alias BAI1 associated protein 2 like 2; BAI1-associated protein 2-like 2; BAI1-associated protein 2-like protein 2; Baiap2l2; BC048937; Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 2; pinkbar; Planar intestinal- and kidney-specific BAR domain protein; RGD1308786; UNQ9336/PRO34007
Gene Symbols BAIAP2L2
Host Species Rabbit
Immunogen A synthesized peptide derived from human BAIAP2L2(Accession Q6UXY1), corresponding to amino acid residues Q120-S170.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207495, 80115
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.