Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAIAP2L2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983076
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3076 Supplier Bioss Supplier No. BS13684R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene binds phosphoinositides and promotes the formation of planar or curved membrane structures. The encoded protein is found in RAB13-positive vesicles and at intercellular contacts with the plasma membrane. [provided by RefSeq, Dec 2012].

Specifications

Antigen BAIAP2L2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BAIAP2L2
Gene Accession No. Q6UXY1
Gene Alias BAI1 associated protein 2 like 2; BAI1-associated protein 2-like 2; BAI1-associated protein 2-like protein 2; Baiap2l2; BC048937; Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 2; Pinkbar; Planar intestinal- and kidney-specific BAR domain protein; UNQ9336/PRO34007
Gene Symbols BAIAP2L2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BAIAP2L2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80115
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.