Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAIAP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 248361AP150UL

Description
BAP3 was initially identified through interaction in a yeast two-hybrid system with the brain-specific angiogenesis inhibitor 1, a p53-target gene that encodes a seven-span transmembrane protein member of the secretin receptor family. BAP3 is predomitly expressed in the brain and possess high homology with Munc13 and synaptotagmin, suggesting that BAP3 may play a role in regulating neurotransmitter release. Recent experiments have shown that BAP3 is induced in certain tumors such as desmoplastic small round cell tumor. Ectopic expression of BAP3 in tumor cells dramatically enhances growth in low serum conditions and colony formation in soft agar, suggesting that the regulated exocytotic pathway may play a role in cancer cell proliferation.Specifications
BAIAP3 | |
Polyclonal | |
Unconjugated | |
BAIAP3 | |
BAI1 associated protein 3, BAIAP3, BAP3, KIAA0734 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
106146145, 545192, 8938 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O94812, Q80TT2 | |
Baiap3 | |
BAIAP3 Fusion Protein Ag21219 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction