Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAK Antibody (38E2), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP174026PECY7
Description
BAK Monoclonal specifically detects BAK in Human samples. It is validated for Flow Cytometry.Specifications
BAK | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
BAK1 | |
Recombinant human BAK1 (29-187aa) purified from E.coli | |
0.1 mL | |
Primary | |
Anti-human BAK1 mAb, clone 8B4, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human BAK1 protein. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
AT38E2 | |
Flow Cytometry | |
Apoptosis regulator BAK, BAK-LIKE, BAKMGC3887, BCL2-antagonist/killer 1, Bcl2-L-7, BCL2L7bcl-2 homologous antagonist/killer, BCL2-like 7 protein, Bcl-2-like protein 7, CDN1, MGC117255, pro-apoptotic protein BAK | |
Mouse | |
Protein G purified | |
Apoptosis, Cancer, Tumor Suppressors | |
578.0 | |
Human | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction