Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAK Monoclonal Antibody (2H9H7)
GREENER_CHOICE

Catalog No. FERMA548385
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548385 Supplier Invitrogen™ Supplier No. MA548385
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BAK Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form oligomers or heterodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein localizes to mitochondria, and functions to induce apoptosis. It interacts with and accelerates the opening of the mitochondrial voltage-dependent anion channel, which leads to a loss in membrane potential and the release of cytochrome c. This protein also interacts with the tumor suppressor P53 after exposure to cell stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAK
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone 2H9H7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene BAK1
Gene Accession No. Q16611
Gene Alias Apoptosis regulator BAK; BAK; Bak1; BAK-LIKE; BCL2 antagonist/killer 1; Bcl2 homologous antagonist/killer; Bcl-2 homologous antagonist/killer; BCL2-antagonist/killer 1; BCL2L7; bcl2-L-7; BCL2-like 7 protein; BCL2-like 7protein; bcl-2-like protein 7; Bcl-2-likeous antagonist/killer; CDN1; MGC117255; MGC3887; N-Bak; N-BAK1; pro-apoptotic protein BAK
Gene Symbols BAK1
Host Species Mouse
Immunogen Purified recombinant fragment of human BAK1 (AA: 29-187) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 578
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.