Learn More
Gibco™ HBSS, calcium, magnesium, phenol red
Description
Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order. Commonly used for processing cells and flushing equipment, sourcing Gibco balanced salt solutions can help streamline your operation by reducing buffer preparation and allowing you to focus on additional critical steps in your biopharmaceutical processes. Discover new outsourcing efficiencies to reclaim time, space, and resources.
Product Features
- Multiple formulations and concentrations available
- Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
- Established products with set specifications are made to order
- Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
- Readily accessible documentation for accelerated qualification
Benefits
- Reduces time spent preparing buffers in-house
- Mitigates space requirement for equipment storage
- Streamlines in-house processes related to formulation, hydration, testing, and validation
- Meets workflow needs across stages of development and manufacturing of biologicals

Specifications
Specifications
Color | Clear, colorless solution |
Form | Liquid |
Product Type | Hanks' Balanced Salt Solution (HBSS) |
Sterility | Sterile-filtered |
With Additives | Calcium, Magnesium, Phenol Red |
Format | Drum |
Manufacturing Quality | cGMP |
Osmolality | 265 to 295 mOsm/kg |
pH | 6.7 to 7.8 |
Quantity | 100 L |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.