Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Balicatib
GREENER_CHOICESDP

Catalog No. 558550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
558550 50 mg
558510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 558550 Supplier Tocris Bioscience™ Supplier No. 5585/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective cathepsin K inhibitor

Balicatib is a potent and selective cathepsin K inhibitor (IC50 values are 1.4, 56 and 480 nM for human, rat and mouse cathepsin K, respectively). Exhibits >300-fold selectivity for cathepsin K over cathepsins L, B and S. Long term supratherapeutic dosing increases tissue levels of cathepsins L and B in rats.

Specifications

Chemical Name or Material N-[1-(Cyanomethylcarbamoyl)cyclohexyl]-4-(4-propylpiperazin-1-yl)benzamide
CAS 354813-19-7
Quantity 50 mg
Target Synthetic Cathepsin Inhibitors
Molecular Formula C23H33N5O2
Formula Weight 411.54
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.