Learn More
GSC Go Science Crazy Ball and Ring Metal Expansion Demonstration
SCI_ED
Catalog No. s13643 Shop All GSC Products
Click to view available options
:
Demonstration; Metal expansion; Ball and ring
Intended for ages 13 and up.
Intended for ages 13 and up.
Includes
Ball; Ring; Instructions
Demonstrates the principles of the expansion of metals. The GSC International Ball and Ring Metal Expansion Demonstration provides a simple and economical visual mechanism for explaining this concept. This demonstration is designed for use in classroom applications.
  • Provides a visible demonstration of the principles of the expansion of metals
  • When the ball is heated it will not pass through the ring, but when cooled the ball passes through the ring freely
  • Ball and ring are mounted on insulated acrylic handles to protect the user's hands from the heat
  • Instructions are included
Specifications
For Use With (Application) Demonstrating the expansion of metals
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.