Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Balloons PROMO
SCI_ED

Catalog No. S04187FB25 Shop All Fisherbrand Products
Click to view available options
Color:
Assorted

Catalog No. S04187FB25

Only null left
Add to Cart
Add to Cart

Commonly used in the science classroom.

  • Dia.: 9 in.
  • Assorted colors

Specifications

Product Type Balloon
Color Assorted
For Use With (Application) For science classroom or with kits
Shape Round
Diameter (English) Outer 9 in.
Diameter (Metric) Outer 23 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.