Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ bamB Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144584
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144584 Supplier Invitrogen™ Supplier No. PA5144584
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

bamB Polyclonal Antibody for Western Blot, ELISA

Part of the outer membrane protein assembly complex (Bam), which is involved in assembly and insertion of beta-barrel proteins into the outer membrane. Nonessential member of the complex, which may orient the flexible periplasmic domain of BamA for interaction with other Bam components, chaperones and nascent outer membrane proteins. Efficient substrate folding and insertion into the outer membrane requires all 5 subunits (PubMed:20378773, PubMed:21823654, PubMed:27686148). A lateral gate may open between the first and last strands of the BamA beta-barrel that allows substrate to insert into the outer membrane; comparison of the structures of complete and nearly complete Bam complexes show there is considerable movement of all 5 proteins (PubMed:27686148, PubMed:26900875, PubMed:26901871).
TRUSTED_SUSTAINABILITY

Specifications

Antigen bamB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.854 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene bamB
Gene Accession No. P77774
Gene Alias yfgL
Gene Symbols bamB
Host Species Rabbit
Immunogen Recombinant E.coli bamB protein (125-386aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 946982
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.