Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BAMBI Monoclonal Antibody (02)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530080
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human BAMBI/NMA.
BAMBI (BMP and activin membrane-bound inhibitor homolog, NMA) is a membrane spanning glycoprotein that acts as a negative regulator of TGF-beta signaling during development. BAMBI transcription regulation is under the influence of beta-catenin, BMP, smad3 and smad4. BAMBI expression can increase in colorectal and hepatocellular carcinomas relative to non-cancerous tissues. BAMBI is expressed at high levels during odontogenesis. BAMBI is coexpressed with Bmp-4 during early Xenopus embryogenesis and can be detected in poorly metastatic human melanoma cell lines.
Specifications
BAMBI | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q13145 | |
BAMBI | |
Recombinant Human BAMBI/NMA Protein (Met1-Ala152). | |
100 μL | |
Primary | |
Human | |
Antibody |
ELISA | |
02 | |
Unconjugated | |
BAMBI | |
2610003H06Rik; BAMBI; BMP and activin membrane bound inhibitor; BMP and activin membrane-bound inhibitor; BMP and activin membrane-bound inhibitor homolog; BMP and activin membrane-bound inhibitor, homolog; kinase-deficient TGFbeta superfamily receptor subunit; NMA; Non-metastatic gene A protein; putative transmembrane protein NMA | |
Mouse | |
Protein A | |
RUO | |
25805 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction