Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAMBI/NMA Antibody (211), FITC , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289698F
Description
BAMBI/NMA Monoclonal antibody specifically detects BAMBI/NMA in Human samples. It is validated for ELISASpecifications
BAMBI/NMA | |
Monoclonal | |
FITC | |
BMP and activin membrane-bound inhibitor homolog, BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), NMANon-metastatic gene A protein, Putative transmembrane protein NMA | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human BAMBI/NMA (NP_036474.1; Met 1-Ala 152). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
211 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
25805 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction