Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAMBI/NMA Antibody (OTI5D5), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271778R
Description
BAMBI/NMA Monoclonal specifically detects BAMBI/NMA in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
BAMBI/NMA | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BAMBI | |
Recombinant protein fragment corresponding to amino acids 21-152 of human BAMBI/NMA (NP_036474) produced in E.coli. | |
0.1 mL | |
25805.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI5D5 | |
Western Blot | |
BMP and activin membrane-bound inhibitor homolog, BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), NMANon-metastatic gene A protein, Putative transmembrane protein NMA | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction