Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAMBI/NMA Antibody (OTI5D5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246563
Description
Ensure accurate, reproducible results in Western Blot
BAMBI/NMA Monoclonal specifically detects BAMBI/NMA in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
BAMBI/NMA | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
BMP and activin membrane-bound inhibitor homolog, BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), NMANon-metastatic gene A protein, Putative transmembrane protein NMA | |
Mouse | |
26.6 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
OTI5D5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
BAMBI | |
Human recombinant protein fragment corresponding to amino acids 21-152 of human BAMBI(NP_036474) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
25805 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction