Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAMBI Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982225
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2225 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2225 Supplier Bioss Supplier No. BS12418R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BAMBI (BMP and activin membrane-bound inhibitor homolog, NMA) is a membrane spanning glycoprotein that acts as a negative regulator of TGF-beta signaling during development. BAMBI transcription regulation is under the influence of beta-catenin, BMP, smad3 and smad4. BAMBI expression can increase in colorectal and hepatocellular carcinomas relative to non-cancerous tissues. BAMBI is expressed at high levels during odontogenesis. BAMBI is coexpressed with Bmp-4 during early Xenopus embryogenesis and can be detected in poorly metastatic human melanoma cell lines.

Specifications

Antigen BAMBI
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BAMBI
Gene Accession No. Q13145
Gene Alias 2610003H06Rik; Bambi; BMP and activin membrane bound inhibitor; BMP and activin membrane-bound inhibitor; BMP and activin membrane-bound inhibitor homolog; BMP and activin membrane-bound inhibitor homolog (Xenopus laevis); BMP and activin membrane-bound inhibitor, homolog; Kinase-deficient TGFbeta superfamily receptor subunit; Nma; Non-metastatic gene A protein; Putative transmembrane protein NMA
Gene Symbols BAMBI
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BAMBI.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25805
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.