Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAMBI Recombinant Rabbit Monoclonal Antibody (211)
GREENER_CHOICE

Catalog No. PIMA530083
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530083 Supplier Invitrogen™ Supplier No. MA530083
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human BAMBI/NMA.

BAMBI (BMP and activin membrane-bound inhibitor homolog, NMA) is a membrane spanning glycoprotein that acts as a negative regulator of TGF-beta signaling during development. BAMBI transcription regulation is under the influence of beta-catenin, BMP, smad3 and smad4. BAMBI expression can increase in colorectal and hepatocellular carcinomas relative to non-cancerous tissues. BAMBI is expressed at high levels during odontogenesis. BAMBI is coexpressed with Bmp-4 during early Xenopus embryogenesis and can be detected in poorly metastatic human melanoma cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAMBI
Applications ELISA
Classification Recombinant Monoclonal
Clone 211
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene BAMBI
Gene Accession No. Q13145
Gene Alias 2610003H06Rik; BAMBI; BMP and activin membrane bound inhibitor; BMP and activin membrane-bound inhibitor; BMP and activin membrane-bound inhibitor homolog; BMP and activin membrane-bound inhibitor, homolog; kinase-deficient TGFbeta superfamily receptor subunit; NMA; Non-metastatic gene A protein; putative transmembrane protein NMA
Gene Symbols BAMBI
Host Species Rabbit
Immunogen Recombinant Human BAMBI/NMA Protein (Met1-Ala152).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25805
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.