Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ bamE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144406
Description
bamE Polyclonal Antibody for Western Blot, ELISA
Part of the outer membrane protein assembly complex (Bam), which is involved in assembly and insertion of beta-barrel proteins into the outer membrane. Nonessential member of the complex that stabilizes the interaction between the essential proteins BamA and BamD. May modulate the conformation of BamA, likely through interactions with BamD. Efficient substrate folding and insertion into the outer membrane requires all 5 subunits (PubMed:20378773, PubMed:21823654, PubMed:27686148). A lateral gate may open between the first and last strands of the BamA beta-barrel that allows substrate to insert into the outer membrane; comparison of the structures of complete and nearly complete Bam complexes show there is considerable movement of all 5 proteins (PubMed:27686148, PubMed:26900875, PubMed:26901871, PubMed:26744406).
Specifications
bamE | |
Polyclonal | |
Unconjugated | |
bamE | |
smpA | |
Rabbit | |
Protein G | |
RUO | |
75205878 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2.67 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P0A937 | |
bamE | |
Recombinant Escherichia coli (strain K12) BAME protein (20-113aa). | |
100 μL | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction