Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BANF1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176750MFV610
Description
BANF1 Polyclonal antibody specifically detects BANF1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ImmunofluorescenceSpecifications
BANF1 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence | |
BAFMGC111161, barrier to autointegration factor 1, barrier-to-autointegration factor, BCRG1, BCRP1, Breakpoint cluster region protein 1, D14S1460 | |
Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of human BANF1. The immunogen is located within the last 50 amino acids of BANF1. Amino Acid Sequence: LKKDEDLFREWLKDT | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
8815 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction