Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BANF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118975
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118975 200 μL
PIPA5122216 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5118975 Supplier Invitrogen™ Supplier No. PA5118975
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BANF1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Barrier-to-autointegration factor 1 (BANF1) is a conserved chromatin protein that non-specifically binds double-stranded DNA. BANF1 also interacts with a family of nuclear proteins that include LAP2, emerin, and MAN1. It is also a host cell component of retroviral pre-integration complexes (PICs), including that of HIV. BANF1 will bind to p55 Gag (the structural precursor of HIV-1 virions) as well as its cleaved product matrix. In addition to being a host cell component of the PIC, it is thought that BANF1 is also present at low levels in incoming virions, and thus might contribute to the assembly or activity of HIV-1 PICs through direct binding to matrix as well as DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BANF1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene BANF1
Gene Accession No. O75531, Q9R1T1
Gene Alias Baf; Banf1; barrier to autointegration factor 1; barrier-to-autointegration factor; Barrier-to-autointegration factor, N-terminally processed; BCRG1; Bcrp1; breakpoint cluster region protein 1; Breakpoint cluster region protein, uterine leiomyoma, 1; Breakpoint cluster region protein, uterine leiomyoma, 1; barrier to autointegration factor; D14S1460; L2bp1; L2bp1/Baf; Lap2 binding protein 1; LAP2-binding protein 1; NGPS
Gene Symbols BANF1
Host Species Rabbit
Immunogen Recombinant protein Barrier To Autointegration Factor 1. The antigen corresponds to amino acid range 2-89 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100622671, 114087, 8815
Target Species Human, Rat, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.