Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BANF1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520328

Catalog No. PIPA520328


Only null left
Add to Cart

Description

Description

Despite its predicted molecular weight, BANF1 often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is human kidney tissue lysate. PA5-20328 can be used with blocking peptide PEP-0448.

Barrier-to-autointegration factor 1 (BANF1) is a conserved chromatin protein that non-specifically binds double-stranded DNA. BANF1 also interacts with a family of nuclear proteins that include LAP2, emerin, and MAN1. It is also a host cell component of retroviral pre-integration complexes (PICs), including that of HIV. BANF1 will bind to p55 Gag (the structural precursor of HIV-1 virions) as well as its cleaved product matrix. In addition to being a host cell component of the PIC, it is thought that BANF1 is also present at low levels in incoming virions, and thus might contribute to the assembly or activity of HIV-1 PICs through direct binding to matrix as well as DNA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BANF1
Polyclonal
Unconjugated
BANF1
Baf; Banf1; barrier to autointegration factor 1; barrier-to-autointegration factor; Barrier-to-autointegration factor, N-terminally processed; BCRG1; Bcrp1; breakpoint cluster region protein 1; Breakpoint cluster region protein, uterine leiomyoma, 1; Breakpoint cluster region protein, uterine leiomyoma, 1; barrier to autointegration factor; D14S1460; L2bp1; L2bp1/Baf; Lap2 binding protein 1; LAP2-binding protein 1; NGPS
Rabbit
Antigen affinity chromatography
RUO
23825, 8815
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O54962, O75531
BANF1
A 15 amino acid peptide from near the carboxy terminus of human BANF1.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.