Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BANF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570404
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 93%.
BANF2 gene ontology annotations related to this gene include DNA binding.
Specifications
BANF2 | |
Polyclonal | |
Unconjugated | |
BANF2 | |
4930517K23Rik; BAF2; BAFL; BAF-L; Baf-like; BANF family member 2; Banf2; barrier to autointegration factor 2; barrier-to-autointegration factor 2; Barrier-to-autointegration factor-like protein; C20orf179; Gm115; likely ortholog of H. sapiens Chromosome 20 open reading frame 179 (C20orf179); LOC538599 protein; RGD1307909; testis secretory sperm-binding protein Li 212a | |
Rabbit | |
Affinity chromatography | |
RUO | |
403171 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8BVR0 | |
BANF2 | |
synthetic peptide directed towards the middle region of human BANF2 (aa 2-51). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction