Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BANF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560062
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560062 Supplier Invitrogen™ Supplier No. PA560062
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLSEPIGEKD VCWVDGISHE LAINLVTKGI NKAYILLGQF LLMHKNEAEF QRWLICCFGA TECEAQQTSH CLKEWCA Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

BANF2 gene ontology annotations related to this gene include DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BANF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BANF2
Gene Accession No. Q9H503
Gene Alias 4930517K23Rik; BAF2; BAFL; BAF-L; Baf-like; BANF family member 2; Banf2; barrier to autointegration factor 2; barrier-to-autointegration factor 2; Barrier-to-autointegration factor-like protein; C20orf179; Gm115; likely ortholog of H. sapiens Chromosome 20 open reading frame 179 (C20orf179); LOC538599 protein; RGD1307909; testis secretory sperm-binding protein Li 212a
Gene Symbols BANF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BANF2. Recombinant protein control fragment (Product #RP-99407).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140836
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.