Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BANP Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079784JF646
Description
BANP Polyclonal antibody specifically detects BANP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
BANP | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BEN domain containing 1, BEND1BEN domain-containing protein 1, BTG3 associated nuclear protein, Btg3-associated nuclear protein, DKFZp761H172, FLJ10177, FLJ20538, protein BANP, SMAR1Scaffold/matrix-associated region-1-binding protein, SMARBP1 | |
The immunogen recognized by this antibody maps to a region between residue 425 and the C-terminus (residue 469) of human BTG3 associated nuclear protein using the numbering given in TrEMBL entry Q96GJ7 (GeneID 54971). | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
54971 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction