Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BANP Monoclonal Antibody (1524CT337.6.32)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537701
Description
BANP Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a protein that binds to matrix attachment regions. The protein forms a complex with p53 and negatively regulates p53 transcription, and functions as a tumor suppressor and cell cycle regulator. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
BANP | |
Monoclonal | |
0.45 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8N9N5 | |
BANP | |
recombinant protein between 30-390 amino acids from human BANP. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot | |
1524CT337.6.32 | |
Unconjugated | |
BANP | |
AA408158; Banp; BEN domain-containing protein 1; BEND1; BTG3 associated nuclear protein; Btg3-associated nuclear protein; Protein BANP; scaffold/matrix-associated region-1-binding protein; Smar1; SMARBP1 | |
Mouse | |
Protein G | |
RUO | |
54971 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction